Learn More
STEMCELL Technologies RoboSep™ Mouse NK Cell Isolation Kit, For processing: 1 x 10^9 cells
SDP
Catalog No. NC1114685
Encompass
The EasySep™ Mouse NK Cell Isolation Kit targets non-NK cells by labeling unwanted cells with biotinylated antibodies and EasySep™ Streptavidin RapidSpheres™, and separates cells without columns using an EasySep™ magnet. Desired cells are simply poured off into a new tube. Isolated cells are immediately available for downstream applications such as flow cytometry, culture or cell-based assays.This product replaces the EasySep™ Mouse NK Cell Enrichment Kit (Catalog #19755) for even faster cell isolations.
Catalog No. NC1114685 Supplier STEMCELL Technologies Supplier No. 19855RF
The EasySep™ Mouse NK Cell Isolation Kit targets non-NK cells by labeling unwanted cells with biotinylated antibodies and EasySep™ Streptavidin RapidSpheres™, and separates cells without columns using an EasySep™ magnet. Desired cells are simply poured off into a new tube. Isolated cells are immediately available for downstream applications such as flow cytometry, culture or cell-based assays.This product replaces the EasySep™ Mouse NK Cell Enrichment Kit (Catalog #19755) for even faster cell isolations.