Learn More
ACROBiosystems ACROBiosystems Mouse NKG2A / CD159a Protein, His Tag
Catalog No. 502106922
Encompass_Preferred
Recombinant Protein;1MG;Mouse NKG2A, His Tag (NKA-M52H5) is expressed from human 293 cells (HEK293). It contains AA Thr 95 - Ile 244 (Accession # Q9Z202-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 19.1kDa. The protein migrates as 33-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6922 Supplier ACROBiosystems Supplier No. NKAM52H51MG
Recombinant Protein;1MG;Mouse NKG2A, His Tag (NKA-M52H5) is expressed from human 293 cells (HEK293). It contains AA Thr 95 - Ile 244 (Accession # Q9Z202-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 19.1kDa. The protein migrates as 33-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.