Learn More
ACROBiosystems Mouse NKG2A CD94 100ug
Catalog No. 502534396
Encompass_Preferred
Mouse NKG2ACD94 Protein His Tag (NC4-M52H9) is expressed from human 293 cells (HEK293). It contains AA Thr 95 - Ile 244 (NKG2A) Ile 32 - Ile 179 (CD94) (Accession # Q9Z202-1 (NKG2A) O54707-1 (CD94)).
Catalog No. 50-253-4396 Supplier ACROBiosystems Supplier No. NC4M52H9100UG
Mouse NKG2ACD94 Protein His Tag (NC4-M52H9) is expressed from human 293 cells (HEK293). It contains AA Thr 95 - Ile 244 (NKG2A) Ile 32 - Ile 179 (CD94) (Accession # Q9Z202-1 (NKG2A) O54707-1 (CD94)).