Learn More
ACROBiosystems Biotinylated Mouse NKG2A & CD94 protein, Fc tag, Avi tag - 25 μg
Catalog No. 502357301
Encompass_Preferred
Recombinant biotinylated heterodimer comprising the extracellular regions of mouse NKG2A and CD94 fused to a human IgG1 Fc and an Avi tag. Expressed in HEK293 cells and supplied lyophilized with trehalose, it is validated for binding ELISA and HABA assay. Supplied as 25 μg.
  • Biotinylated by site-specific enzymatic labeling for consistent probe orientation.
  • Fc tag enables dimerization and capture in immunoassays.
  • Mammalian expression preserves native glycosylation.
  • Covers residues Thr 95-Ile 244 (NKG2A) and Ile 32-Ile 179 (CD94).
  • Calculated molecular weight 63.0 kDa; migrates ~80-90 kDa when glycosylated.
  • Purity greater than 90% by SDS-PAGE.
  • Lyophilized formulation with trehalose for extended stability; avoid repeated freeze-thaw cycles.
  • Validated for binding ELISA and HABA assay.
Catalog No. 50-235-7301 Supplier ACROBiosystems Supplier No. NC4M82F525UG
Recombinant biotinylated heterodimer comprising the extracellular regions of mouse NKG2A and CD94 fused to a human IgG1 Fc and an Avi tag. Expressed in HEK293 cells and supplied lyophilized with trehalose, it is validated for binding ELISA and HABA assay. Supplied as 25 μg.
  • Biotinylated by site-specific enzymatic labeling for consistent probe orientation.
  • Fc tag enables dimerization and capture in immunoassays.
  • Mammalian expression preserves native glycosylation.
  • Covers residues Thr 95-Ile 244 (NKG2A) and Ile 32-Ile 179 (CD94).
  • Calculated molecular weight 63.0 kDa; migrates ~80-90 kDa when glycosylated.
  • Purity greater than 90% by SDS-PAGE.
  • Lyophilized formulation with trehalose for extended stability; avoid repeated freeze-thaw cycles.
  • Validated for binding ELISA and HABA assay.