Learn More
Raybiotech Inc Mouse NKG2D ELISA Kit
Supplier:  Raybiotech Inc ELMNKG2D1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 1.8 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Mouse NKG2D.
Catalog No. 50-195-0099