Learn More
Biomatik Corporation Mouse NLR Family, Pyrin Domain Containing Protein 3 (NLRP3) ELISA Kit
Supplier:  Biomatik Corporation EKU12250
Encompass_Preferred
Mouse NLR Family, Pyrin Domain Containing Protein 3 (NLRP3) ELISA Kit for detecting NLRP3 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: T1W2H6. Gene ID: 216799. Target Synonyms: AVP; AGTAVPRL; AII; AII/AVP; C1orf7; CIAS1; CLR1.1; FCAS; FCU; MWS; NALP3; PYPAF1; Cryopyrin; Cold Induced Autoinflammatory Syndrome 1; Caterpiller-Like Receptor 1.1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7530