Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Mouse Noggin, Animal-Free Recombinant Protein, PeproTech®

Catalog No. 501628512
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 μg
5 μg
50 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-162-8512 250 μg
AF2503850UG 50 μg
AF250385UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-162-8512 Supplier Gibco™ Supplier No. AF25038250UG
Only null left
Add to Cart
Add to Cart

Recombinant Protein

AF-250-38-1MG will be provided as 2 x 500 μg (AF-250-38-500UG). Recombinant Murine Noggin is a 46.4 kDa disulfide-linked homodimer consisting of two 206 amino acid polypeptide chains. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Noggin is involved in numerous developmental processes, such as neural tube fusion and joint formation. The morphogenesis of organs is initiated by a downgrowth from a layer of epithelial stem cells. This process is achieved through the receipt of signals from 1) a WNT protein (WNT3A) to stabilize beta-catenin; and 2) Noggin, which is a bone morphogenetic protein inhibitor. Noggin mutations in unrelated families with proximal symphalangism (SYM1) and multiple synostoses syndrome (SYNS1) have been identified, which have multiple joint fusion as their principal defect.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P97466
For Use With (Application) Functional Assay
Format Lyophilized
Formulation protein with no preservative
Gene ID (Entrez) 18121
Molecular Weight (g/mol) 46.4 kDa
Name Mouse Noggin, Animal-Free
Purification Method HPLC, SDS-PAGE
Quantity 250 μg
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/ μg
Gene Alias NOG; Noggin; Noggin precursor; noggin; LOW QUALITY PROTEIN: noggin; SYM1; symphalangism 1 (proximal); SYNS1; SYNS1A
Common Name Noggin
Gene Symbol NOG
Biological Activity Determined by its ability to inhibit 5.0 ng/ml of BMP-4 induced alkaline phosphatase production by ATDC5 chondrogenic cells. The expected ED50 for this effect is 1.0-2.0 ng/ml of Noggin.
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence MQHYLHIRPA PSDNLPLVDL IEHPDPIFDP KEKDLNETLL RSLLGGHYDP GFMATSPPED RPGGGGGPAG GAEDLAELDQ LLRQRPSGAM PSEIKGLEFS EGLAQGKKQR LSKKLRRKLQ MWLWSQTFCP VLYAWNDLGS RFWPRYVKVG SCFSKRSCSV PEGMVCKPSK SVHLTVLRWR CQRRGGQRCG WIPIQYPIIS ECKCSC
Content And Storage -20°C
Activity Determined by its ability to inhibit 5.0 ng/ml of BMP-4 induced alkaline phosphatase production by ATDC5 chondrogenic cells. The expected ED50 for this effect is 1.0-2.0 ng/ml of Noggin.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.