Learn More
Euthanex Mouse Nosecone
Catalog No. 501540155
Encompass_Preferred
Mouse Nosecones are custom designed for small animals, ensure a tight fit and minimal dead space. Nosecones include rubber plugs to close-off gas supply when not in use.
Catalog No. 50-154-0155 Supplier Euthanex Supplier No. EZ110
Mouse Nosecones are custom designed for small animals, ensure a tight fit and minimal dead space. Nosecones include rubber plugs to close-off gas supply when not in use.