Learn More
Braintree Scientific EZ Anesthesia Nose Cone for mice w/black plug
SDP
Catalog No. NC9904989
Encompass
E-Z Anesthesia nosecones are custom-designed for small animals, ensuring a tight fit and minimal dead space. They are available in three sizes to fit mice, rats and rabbits. Nosecones include rubber plugs to close off gas supply when not in use. Nosecone plugs are also available separately. **Sold for research purposes only and not for human use**
Catalog No. NC9904989 Supplier Braintree Scientific Supplier No. EZ110
 Restricted Item
Add to Cart
Add to Cart
E-Z Anesthesia nosecones are custom-designed for small animals, ensuring a tight fit and minimal dead space. They are available in three sizes to fit mice, rats and rabbits. Nosecones include rubber plugs to close off gas supply when not in use. Nosecone plugs are also available separately. **Sold for research purposes only and not for human use**