Learn More
Vetequip Inc MOUSE NOSECONE
SDP
Catalog No. NC9854822
Encompass_Preferred
This item has a minimum qty of 5 per supplier requirements.
Cone; Nose; Rodent-Mouse; 9mm; Approximately 3/4in. L; funnel-shaped cone that makes an excellent anesthesia mask by cutting down on in-mask dead space without restricting access to the eyes or cranium
Catalog No. NC9854822 Supplier Vetequip Inc Supplier No. 921609
This item has a minimum qty of 5 per supplier requirements.
Cone; Nose; Rodent-Mouse; 9mm; Approximately 3/4in. L; funnel-shaped cone that makes an excellent anesthesia mask by cutting down on in-mask dead space without restricting access to the eyes or cranium