Learn More
BIOHIPPO Mouse NPY1R Microsample
SDP
Catalog No. 5000313984
Encompass
Mouse NPY1R Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of NPY1R in tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 059 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse NPY1R Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse NPY1R Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse NPY1R biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13984 Supplier BIOHIPPO Supplier No. BHE15211084
Mouse NPY1R Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of NPY1R in tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 059 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse NPY1R Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse NPY1R Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse NPY1R biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color