Learn More
Sino Biological Mouse NR1H4 Protein 500ug (Sino Biological)
Supplier:  Sino Biological 53744M07E500
Encompass_Preferred
A DNA sequence encoding the Mouse NR1H4 (BC015261.1) (Thr255−Gln484) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-237-3206