Learn More
Sino Biological Anti-5'-nucleotidase/CD73/NT5E Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50231RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse NT5E (rM NT5E; Catalog#50231-M08H; NP_035981.1; Met 1-Lys 549). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-9561