Learn More
Cayman Chemical Mouse ntI-6HIS IgG HRP 100ug
SDP
Catalog No. 502966207
Encompass_Preferred
Horseradish Peroxidase conjugated to Mouse Anti-6Histidine Antibody: Mouse monoclonal anti-6x histidine IgG, clone AD1.1.10 Enzyme: Horseradish Peroxidase Uses: ELISA, microscopy, Western blot detection. Recommended dilution of 1:500-1:10,000. Starting dilution: 1:2,000.
Catalog No. 50-296-6207 Supplier Cayman Chemical Supplier No. 16745100 UG
Horseradish Peroxidase conjugated to Mouse Anti-6Histidine Antibody: Mouse monoclonal anti-6x histidine IgG, clone AD1.1.10 Enzyme: Horseradish Peroxidase Uses: ELISA, microscopy, Western blot detection. Recommended dilution of 1:500-1:10,000. Starting dilution: 1:2,000.