Learn More
Cayman Chemical Mouse ntI6HIS IgG DyLIght 1ea
SDP
Catalog No. 502951464
Encompass_Preferred
Mouse anti-6His (6x Histidine) IgG (Clone AD1.1.10) conjugated to DyLight 488 Antibody: Mouse monoclonal anti-6His IgG Clone: AD1.1.10 Isotype: IgG Dye: DyLight 488 Excitation max. λ: 493 nm Emission max. λ: 518 nm Uses: Flow cytometry, fluorescence-activated cell sorting, and microscopy
Catalog No. 50-295-1464 Supplier Cayman Chemical Supplier No. 112461 EA
Mouse anti-6His (6x Histidine) IgG (Clone AD1.1.10) conjugated to DyLight 488 Antibody: Mouse monoclonal anti-6His IgG Clone: AD1.1.10 Isotype: IgG Dye: DyLight 488 Excitation max. λ: 493 nm Emission max. λ: 518 nm Uses: Flow cytometry, fluorescence-activated cell sorting, and microscopy