Learn More
Cayman Chemical Mouse ntIBIotIn IgG SureLIg
SDP
Catalog No. 502967733
Encompass_Preferred
Mouse Anti-Biotin IgG1 (Clone B098M) conjugated to SureLight™ PE Antibody: Mouse Anti-Biotin IgG1 Dye: SureLight R-phycoerythrin Excitation max. λ: 565>498 nm Emission max. λ: 578 nm Clone Number: B098M Isotype: IgG1 Uses: Flow cytometry, fluorescence-activated cell sorting, and protein microarrays
Catalog No. 50-296-7733 Supplier Cayman Chemical Supplier No. 16694100 UG
Mouse Anti-Biotin IgG1 (Clone B098M) conjugated to SureLight™ PE Antibody: Mouse Anti-Biotin IgG1 Dye: SureLight R-phycoerythrin Excitation max. λ: 565>498 nm Emission max. λ: 578 nm Clone Number: B098M Isotype: IgG1 Uses: Flow cytometry, fluorescence-activated cell sorting, and protein microarrays