Learn More
Cayman Chemical Mouse ntIcmyc IgG AlkalIn 1ea
SDP
Catalog No. 502955609
Encompass_Preferred
For immunochemical detection of His-tagged proteins
Catalog No. 50-295-5609 Supplier Cayman Chemical Supplier No. 134631 EA
For immunochemical detection of His-tagged proteins