Learn More
Cayman Chemical Mouse ntIcmyc IgG conjugat 1ea
SDP
Catalog No. 502951415
Encompass_Preferred
For direct immunofluorescent detection of c-myc fusion proteins
Catalog No. 50-295-1415 Supplier Cayman Chemical Supplier No. 111161 EA
For direct immunofluorescent detection of c-myc fusion proteins