Learn More
Cayman Chemical Mouse ntIcmyc IgG EuW1024 20ug
SDP
Catalog No. 502967290
Encompass_Preferred
Antibody: Mouse mononcolonal anti-c-myc IgG (clone 9E10) Dye: Europium Chelate W-1024 Excitation max. λ: 340 nm Emission max. λ: 615 nm Uses: TR-FRET and High throughput screening (HTS) applications Specificity: Recognizes the 9E10 epitope (EQKLISEEDL sequence) derived from the human c-myc protein.
Catalog No. 50-296-7290 Supplier Cayman Chemical Supplier No. 1657720 UG
Antibody: Mouse mononcolonal anti-c-myc IgG (clone 9E10) Dye: Europium Chelate W-1024 Excitation max. λ: 340 nm Emission max. λ: 615 nm Uses: TR-FRET and High throughput screening (HTS) applications Specificity: Recognizes the 9E10 epitope (EQKLISEEDL sequence) derived from the human c-myc protein.