Learn More
Cayman Chemical Mouse ntIcmyc IgG SureLIgh
SDP
Catalog No. 502967354
Encompass_Preferred
Mouse anti-c-myc IgG clone 9E10 (EQKLISEEDL) conjugated to SureLight™ APC Antibody: Mouse monoclonal anti-c-myc IgG Clone: 9E10 (EQKLISEEDL) Dye: SureLight™ Allophycocyanin (APC) Excitation max. λ: 652 nm Emission max. λ: 657.5 nm Uses: Flow cytometry, high throughput screening (HTS), and fluorescence resonance energy transfer (FRET) assays
Catalog No. 50-296-7354 Supplier Cayman Chemical Supplier No. 16602100 UG
Mouse anti-c-myc IgG clone 9E10 (EQKLISEEDL) conjugated to SureLight™ APC Antibody: Mouse monoclonal anti-c-myc IgG Clone: 9E10 (EQKLISEEDL) Dye: SureLight™ Allophycocyanin (APC) Excitation max. λ: 652 nm Emission max. λ: 657.5 nm Uses: Flow cytometry, high throughput screening (HTS), and fluorescence resonance energy transfer (FRET) assays