Learn More
Cayman Chemical Mouse ntIDYKDDDDKClon M2 100ug
SDP
Catalog No. 502937048
Encompass_Preferred
Horseradish Peroxidase conjugated to Mouse Anti-DYKDDDDK Antibody: Mouse monoclonal anti-DYKDDDDK (Clone M2) Enzyme: Horseradish Peroxidase Uses: Western blot detection. Recommended dilution of 1:500-1:10,000. Starting dilution: 1:2,000.
Catalog No. 50-293-7048 Supplier Cayman Chemical Supplier No. 27096100 UG
Horseradish Peroxidase conjugated to Mouse Anti-DYKDDDDK Antibody: Mouse monoclonal anti-DYKDDDDK (Clone M2) Enzyme: Horseradish Peroxidase Uses: Western blot detection. Recommended dilution of 1:500-1:10,000. Starting dilution: 1:2,000.