Learn More
Cayman Chemical Mouse ntIHA IgG DyLIght 65
SDP
Catalog No. 502951581
Encompass_Preferred
Mouse anti-HA (Hemagglutinin) IgG1 (Clone 16B12) conjugated to DyLight 650 Antibody: Mouse anti-HA IgG (clone 16B12) Dye: DyLight 650 Reactivity: Human influenza HA tagged proteins Uses: Fluorescence-activated cell sorting, flow cytometry, and microscopy
Catalog No. 50-295-1581 Supplier Cayman Chemical Supplier No. 11266100 UG
Mouse anti-HA (Hemagglutinin) IgG1 (Clone 16B12) conjugated to DyLight 650 Antibody: Mouse anti-HA IgG (clone 16B12) Dye: DyLight 650 Reactivity: Human influenza HA tagged proteins Uses: Fluorescence-activated cell sorting, flow cytometry, and microscopy