Learn More
Sino Biological Anti-TrkA Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 51103RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse TrkA / NTRK1 (Catalog#51103-M08H; NP_001028296.1; Met1-Gly420). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1789