Learn More
Sino Biological Recombinant Mouse TrkA Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 51103M08H1
Encompass_Preferred
A DNA sequence encoding the mouse NTRK1 (NP_001028296.1)(Met1-Gly420) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-255-3010