Learn More
Sino Biological Recombinant Anti-TrkC Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50320R101100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse TrkC / NTRK3 (rM TrkC / NTRK3; Catalog#50320-M08H; NP_032772.3; Met1-Thr429).
Catalog No. 50-255-3311