Learn More
Sino Biological Anti-TrkC Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50320RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse TrkC / NTRK3 extracellular domain (rM TrkC; Catalog#50320-M08H; NP_032772.3; Met 1-Thr 429). TrkC specific IgG was purified by mouse TrkC affinity chromatography.
Catalog No. 50-255-3309