Learn More
American Research Products Inc Mouse Nuclear factor erythroid
Catalog No. 5000326652
Encompass
Mouse Nuclear factor erythroid 2-related factor 2 (Nrf2) ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma cell culture supernates Method Sandwich
Catalog No. 50-003-26652 Supplier American Research Products Inc Supplier No. CSBE16188M
Mouse Nuclear factor erythroid 2-related factor 2 (Nrf2) ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma cell culture supernates Method Sandwich