Learn More
ACROBiosystems ACROBiosystems Mouse Nucleolin / NCL Protein, Mouse IgG2a Fc Tag
Supplier:  ACROBiosystems NULM52531MG
Encompass_Preferred
Recombinant Protein;1MG;Mouse Nucleolin, Mouse IgG2a Fc Tag (NUL-M5253) is expressed from human 293 cells (HEK293). It contains AA Val 2 - Glu 707 (Accession # P09405-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 103.5 kDa. The protein migrates as 130-150 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6835