Learn More
BIOHIPPO Mouse NUP62 Microsample
SDP
Catalog No. 5000314410
Encompass
Mouse NUP62 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of NUP62 in Tissue homogenates and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 34 3 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse NUP62 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse NUP62 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse NUP62 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14410 Supplier BIOHIPPO Supplier No. BHE15212409
Mouse NUP62 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of NUP62 in Tissue homogenates and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 34 3 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse NUP62 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse NUP62 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse NUP62 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color