Learn More
Biomatik Corporation Mouse Odorant Binding Protein 2A (OBP2A) ELISA Kit
Supplier:  Biomatik Corporation EKU09132
Encompass_Preferred
Mouse Odorant Binding Protein 2A (OBP2A) ELISA Kit for detecting OBP2A in tissue homogenates, cell lysates, saliva, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.221ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8K1H9. Gene ID: 227627. Target Synonyms: OBP2-A; OBP; OBP2C; OBPIIa; hOBPIIa. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8563