Learn More
Diagnostic Biosystems MOUSE ON MOUSE MOM UNOVUE HRP MOUSE ON MOUSE MOM UNOVUE HRP
Supplier: Diagnostic Biosystems MMU25PDR
Mouse on Mouse (MOM) Blocking Reagent is a unique serum-free blocking solution used to reduce nonspecific background staining from mouse antibodies. This blocking reagent has been shown to be more superior than serum-based blocking solutions.\n\nBlock nonspecific background staining observed during immunostaining using mouse antibodies on mouse tissue.