Learn More
Biomatik Corporation Mouse Optic Atrophy 1, Autosomal Dominant (OPA1) ELISA Kit
Supplier:  Biomatik Corporation EKU12225
Encompass_Preferred
Mouse Optic Atrophy 1, Autosomal Dominant (OPA1) ELISA Kit for detecting OPA1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P58281. Gene ID: 74143. Target Synonyms: MGM1; NPG; NTG; largeG; Dynamin-Like 120 kDa Protein, Mitochondrial; Optic atrophy protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6056