Learn More
Biomatik Corporation Mouse Ornithine Transcarbamylase (OTC) ELISA Kit
Supplier:  Biomatik Corporation EKU12175
Encompass_Preferred
Mouse Ornithine Transcarbamylase (OTC) ELISA Kit for detecting OTC in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P11725. Gene ID: 18416. Target Synonyms: OCTD; OCT; OTCase; Ornithine Transcarbamylase; Ornithine Carbamoyltransferase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6459