Learn More
Biomatik Corporation Mouse Orosomucoid 2 (ORM2) ELISA Kit
Supplier:  Biomatik Corporation EKU11322
Encompass_Preferred
Mouse Orosomucoid 2 (ORM2) ELISA Kit for detecting ORM2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 9.4-600ng/ml. Sensitivity: 3.7ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P07361. Gene ID: 18406. Target Synonyms: AGP-B; AGP2; Alpha-1-Acid Glycoprotein Type 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7442