Learn More
Sino Biological Recombinant Mouse Oncostatin M/OSM Protein
Supplier:  Sino Biological 50112MNAE1
Encompass_Preferred
A DNA sequence encoding the mouse OSM (P53347) (Ala24-Arg206) was expressed with an initial Met at the N-terminal.
Catalog No. 50-255-3455