Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse Osteoactivin ELISA Kit

Catalog No. EM59RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EM59RB 96 Tests
EM59RBX5 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EM59RB Supplier Invitrogen™ Supplier No. EM59RB
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

Mouse Osteoactivin quantitates mouse Osteoactivin in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant mouse Osteoactivin.

The protein encoded by this gene is a type I transmembrane glycoprotein which shows homology to the pMEL17 precursor, a melanocyte-specific protein. GPNMB shows expression in the lowly metastatic human melanoma cell lines and xenografts but does not show expression in the highly metastatic cell lines. GPNMB may be involved in growth delay and reduction of metastatic potential. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q99P91
Assay Range 80-20000 pg/mL
Assay Sensitivity 80 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Mouse
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias Dchil; DC-HIL; dendritic cell-associated heparin sulfate proteoglycan-dependent integrin ligand; dendritic cell-associated transmembrane protein; glycoprotein (transmembrane) nmb; glycoprotein nmb; glycoprotein nmb-like protein; glycoprotein nonmetastatic melanoma protein B; Gpnmb; hematopoietic growth factor inducible neurokinin-1; Hematopoietic growth factor inducible neurokinin-1 type; Hgfin; ipd; iris pigment dispersion; NMB; Osteoactivin; si:ch211-155k24.6; transmembrane glycoprotein HGFIN; transmembrane glycoprotein NMB; Unknown (protein for MGC:133733); UNQ1725/PRO9925Dchil; DC-HIL; dendritic cell-associated heparin sulfate proteoglycan-dependent integrin ligand; dendritic cell-associated transmembrane protein; glycoprotein (transmembrane) nmb; glycoprotein nmb; glycoprotein nmb-like protein; glycoprotein nonmetastatic melanoma protein B; Gpnmb; hematopoietic growth factor inducible neurokinin-1; Hematopoietic growth factor inducible neurokinin-1 type; Hgfin; ipd; iris pigment dispersion; NMB; Osteoactivin; si:ch211-155k24.6; transmembrane glycoprotein HGFIN; transmembrane glycoprotein NMB; Unknown (protein for MGC:133733); UNQ1725/PRO9925;
Gene ID (Entrez) 93695
Gene Symbol GPNMB
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target GPNMB
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.