Learn More
American Research Products Inc Mouse Osteocalcin Bone gla pro
Catalog No. 5000326283
Encompass
Mouse Osteocalcin/Bone gla protein OT/BGP ELISA kit from CUSABIO Detection Range 78 pg/ml - 5000 pg/ml Sensitivity 19 5 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26283 Supplier American Research Products Inc Supplier No. CSBE06917M
Mouse Osteocalcin/Bone gla protein OT/BGP ELISA kit from CUSABIO Detection Range 78 pg/ml - 5000 pg/ml Sensitivity 19 5 pg/ml Sample serum plasma tissue homogenates Method Sandwich