Learn More
Biomatik Corporation Mouse Osteocalcin (OC) Instant ELISA Kit
Supplier:  Biomatik Corporation EKU11893
Encompass_Preferred
Mouse Osteocalcin (OC) Instant ELISA Kit for detecting OC in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 1h, 10min. Accession Number: P86546. Gene ID: 12096. Target Synonyms: BGLAP; OT; BGP; Bone Gla Protein; Bone Gamma-Carboxyglutamate Protein; Gamma-carboxyglutamic acid-containing protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8068