Learn More
American Research Products Inc Mouse Osteoclast associated im
Catalog No. 5000328423
Encompass
Mouse Osteoclast-associated immunoglobulin-like receptor (OSCAR) ELISA kit from CUSABIO Detection Range 37 5 pg/mL-2400 pg/mL Sensitivity 9 375 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28423 Supplier American Research Products Inc Supplier No. CSBEL017255MO
Mouse Osteoclast-associated immunoglobulin-like receptor (OSCAR) ELISA kit from CUSABIO Detection Range 37 5 pg/mL-2400 pg/mL Sensitivity 9 375 pg/mL Sample serum plasma tissue homogenates Method Sandwich