Learn More
Biomatik Corporation Mouse Osterix (OSX) ELISA Kit
Supplier:  Biomatik Corporation EKU06440
Encompass_Preferred
Mouse Osterix (OSX) ELISA Kit for detecting OSX in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8VI67. Gene ID: 170574. Target Synonyms: Sp7 Transcription Factor; Specificity Protein 7; Zinc finger protein osterix. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6411