Learn More
American Research Products Inc Mouse oValbumin specific IgE
Supplier:  American Research Products Inc CSBE08914MIS
Encompass
Mouse oValbumin specific IgE OVA sIgE ELISA Kit from CUSABIO Detection Range 0 24ng/mL-15ng/mL Sensitivity 0 12ng/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-30087