Learn More
ACROBiosystems ACROBiosystems Mouse OX40 Ligand / TNFSF4 Protein, His Tag
Supplier:  ACROBiosystems TN4M5241500UG
Encompass_Preferred
Recombinant Protein;500UG;Mouse OX40 Ligand, His Tag (TN4-M5241) is expressed from human 293 cells (HEK293). It contains AA Gln 49 - Leu 198 (Accession # P43488-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 18.3 kDa. The protein migrates as 20 kDa and 22-25 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse OX40 Ligand, His Tag (Cat. No. TN4-M5241) at 0.5 μg/mL (100 μL/well) can bind Mouse OX40, Fc Tag (Cat. No. OX0-M5259) with a linear range of 10-16 ng/mL (QC tested).
Catalog No. 50-210-6743