Learn More
American Research Products Inc Mouse oXidized high density li
Catalog No. 5000328292
Encompass
Mouse oXidized high density lipoprotein (OX-HDL) ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-28292 Supplier American Research Products Inc Supplier No. CSBEQ027598MO
Mouse oXidized high density lipoprotein (OX-HDL) ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma tissue homogenates Method Competitive