Learn More
Biomatik Corporation Mouse Oxidized Low Density Lipoprotein (OxLDL) CLIA Kit
Supplier:  Biomatik Corporation EKU09951
Encompass_Preferred
Mouse Oxidized Low Density Lipoprotein (OxLDL) CLIA Kit for detecting OxLDL in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.54pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Target Synonyms: OL. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7451