Learn More
American Research Products Inc Mouse oXidized low density lip
Catalog No. 5000328928
Encompass
Mouse oXidized low density lipoprotein OXLDL ELISA Kit from CUSABIO Detection Range 0 45 nmol/mL-30 nmol/mL Sensitivity 0 11 nmol/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28928 Supplier American Research Products Inc Supplier No. CSBE07933M
Mouse oXidized low density lipoprotein OXLDL ELISA Kit from CUSABIO Detection Range 0 45 nmol/mL-30 nmol/mL Sensitivity 0 11 nmol/mL Sample serum plasma tissue homogenates Method Sandwich