Learn More
Biomatik Corporation Mouse Oxidized Low Density Lipoprotein (OxLDL) Wide-range ELISA Kit
Supplier:  Biomatik Corporation EKU10567
Encompass_Preferred
Mouse Oxidized Low Density Lipoprotein (OxLDL) Wide-range ELISA Kit for detecting OxLDL in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.12-200ng/ml. Sensitivity: 1.34ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Target Synonyms: OL. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8329