Learn More
BIOHIPPO Mouse OxLDLR Microsample
SDP
Catalog No. 5000313710
Encompass
Mouse OxLDLR Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of OxLDLR in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 1 57-100 ng/mL with a sensitivity of 0 48 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse OxLDLR Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse OxLDLR Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse OxLDLR biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13710 Supplier BIOHIPPO Supplier No. BHE15212474
Mouse OxLDLR Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of OxLDLR in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 1 57-100 ng/mL with a sensitivity of 0 48 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse OxLDLR Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse OxLDLR Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse OxLDLR biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color