Learn More
Raybiotech Inc Mouse PAI-1 ELISA Kit
Supplier:  Raybiotech Inc ELMPAII1
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 80 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects mouse and human PAI-1.
Catalog No. 50-195-0106