Learn More
Abcam Recombinant Mouse PAI1 protein, 10UG
Supplier:  Abcam AB18586210UG
Encompass_Preferred
Recombinant Mouse PAI1 protein is a Mouse Full Length protein, in the 24 to 402 aa range, expressed in Mammalian, with >95% purity, < 1 EU/µg endotoxin level and suitable for SDS-PAGE, HPLC.
Protein/ Peptide/ Enzyme Synonyms: Mr1, Pai1, Planh1, Serpine1, Plasminogen activator inhibitor 1, PAI, PAI-1, Endothelial plasminogen activator inhibitor, Serpin E1
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2339911