Learn More
American Research Products Inc Mouse pancreatic alpha amylase
Catalog No. 5000327595
Encompass
Mouse pancreatic alpha amylase (AMY2A) ELISA kit from CUSABIO Detection Range 9 38 mU/mL-600 mU/mL Sensitivity 2 3 mU/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-27595 Supplier American Research Products Inc Supplier No. CSBEL001689MO
Mouse pancreatic alpha amylase (AMY2A) ELISA kit from CUSABIO Detection Range 9 38 mU/mL-600 mU/mL Sensitivity 2 3 mU/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich